Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,6-Anhydro-β-D-mannopyranose | 14168-65-1 | MFCD00069698 | 99.2% | 162.14 | C6H10O5 | 250MG
SDP

Catalog No. 5000236772
Encompass_Preferred

Mannosan (1,6-anhydro-β-D-mannopyranose) is an anhydro sugar reagent supplied as a white to off-white solid for use in glycobiology and carbohydrate research. It is provided in multiple laboratory pack sizes for analytical and preparative work, and has formula C6H10O5 and molecular weight 162.14.

  • Purity determined at 99.18%.
  • Molecular formula C6H10O5 and molecular weight 162.14.
  • Available in small laboratory pack sizes suitable for research use.
  • White to off-white solid, easy to handle and store.
  • Used as a reagent in glycobiology and carbohydrate chemistry applications.

Catalog No. 50-002-36772 Supplier Medchemexpress LLC Supplier No. HY124095250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mannosan (1,6-anhydro-β-D-mannopyranose) is an anhydro sugar reagent supplied as a white to off-white solid for use in glycobiology and carbohydrate research. It is provided in multiple laboratory pack sizes for analytical and preparative work, and has formula C6H10O5 and molecular weight 162.14.

  • Purity determined at 99.18%.
  • Molecular formula C6H10O5 and molecular weight 162.14.
  • Available in small laboratory pack sizes suitable for research use.
  • White to off-white solid, easy to handle and store.
  • Used as a reagent in glycobiology and carbohydrate chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.