Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane 90.0+%, TCI America™
SDP

Catalog No. B23405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23405G Supplier TCI America Supplier No. B23405G

Specifications

CAS 2264-01-9
Color Yellow
Molecular Formula C12H10F8O4
MDL Number MFCD00078412
UN Number 3532
Quantity 5 g
Synonym 2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
InChI Key BWTMTZBMAGYMOD-UHFFFAOYSA-N
SMILES C=CC(=O)OCC(C(C(C(COC(=O)C=C)(F)F)(F)F)(F)F)(F)F
IUPAC Name (2,2,3,3,4,4,5,5-octafluoro-6-prop-2-enoyloxyhexyl) prop-2-enoate
Molecular Weight (g/mol) 370.20
PubChem CID 2775863
Formula Weight 370.20
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.