Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,6-bis(cyclohexyloximinocarbonyl)hexane | 83654-05-1 | MFCD00210844 | 99.4% | 394.51 g·mol⁻1 | C20H34N4O4 | 50 MG
SDP

Catalog No. 5000239834
Encompass_Preferred

RHC 80267 is a small-molecule inhibitor of diacylglycerol lipase (DAGL) used as a research reagent to probe lipid-signaling pathways. It exhibits inhibitory activity in platelets and mammalian cells and may affect related enzymes in some assays. Supplied as a solid for biochemical and cellular studies, it is intended for research use only.

  • Potent DAGL inhibitor with reported activity in biological assays.
  • High purity: 99.4%.
  • Chemical formula: C20H34N4O4.
  • Molecular weight: 394.51 g·mol⁻1.
  • Solid form suitable for dissolution and formulation in organic solvents.
  • Common applications include biochemical assays and cellular studies.

Catalog No. 50-002-39834 Supplier Medchemexpress LLC Supplier No. HY10741650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RHC 80267 is a small-molecule inhibitor of diacylglycerol lipase (DAGL) used as a research reagent to probe lipid-signaling pathways. It exhibits inhibitory activity in platelets and mammalian cells and may affect related enzymes in some assays. Supplied as a solid for biochemical and cellular studies, it is intended for research use only.

  • Potent DAGL inhibitor with reported activity in biological assays.
  • High purity: 99.4%.
  • Chemical formula: C20H34N4O4.
  • Molecular weight: 394.51 g·mol⁻1.
  • Solid form suitable for dissolution and formulation in organic solvents.
  • Common applications include biochemical assays and cellular studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.