Learn More
Medchemexpress LLC 1,6-Di-O-galloyl-β-D-glucose | 23363-08-8 | MFCD00066058 | 90.0% | 484.36 g/mol | C20H20O14 | 1mg
SDP
Catalog No. 5000269899
Encompass_Preferred
1 6-Di-O-galloyl- -D-glucose is a compound found in the fruit of Phyllanthus emblica 1 6-Di-O-galloyl- -D-glucose has HIV-1 reverse transcriptase (RT) inhibitory activity with an IC50 value of 270 M The inhibitory mechanism of 1 6-Di-O-galloyl- -D-glucose is competitive inhibition of the template primer and non-competitive inhibition of the substrate (dTTP) 1 6-Di-O-galloyl- -D-glucose can be used in anti-HIV research[1]
Catalog No. 50-002-69899 Supplier Medchemexpress LLC Supplier No. HYW7190411MG
1 6-Di-O-galloyl- -D-glucose is a compound found in the fruit of Phyllanthus emblica 1 6-Di-O-galloyl- -D-glucose has HIV-1 reverse transcriptase (RT) inhibitory activity with an IC50 value of 270 M The inhibitory mechanism of 1 6-Di-O-galloyl- -D-glucose is competitive inhibition of the template primer and non-competitive inhibition of the substrate (dTTP) 1 6-Di-O-galloyl- -D-glucose can be used in anti-HIV research[1]