Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Dibromo-2-naphthol 97.0+%, TCI America™
SDP

Catalog No. D187225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187225G 25 g
D1872250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D187225G Supplier TCI America Supplier No. D187225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16239-18-2
Molecular Formula C10H6Br2O
Molecular Weight (g/mol) 301.97
MDL Number MFCD00003870
InChI Key VKESFYLPKHQOOA-UHFFFAOYSA-N
Synonym 1,6-dibromo-2-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-beta-naphthol, 1,6-dibromo-.beta.-naphthol, pubchem22920, 2-naphthol,6-dibromo, 1,6 dibromo-2-napthol, acmc-1c8jh
PubChem CID 27768
IUPAC Name 1,6-dibromonaphthalen-2-ol
SMILES OC1=CC=C2C=C(Br)C=CC2=C1Br

Specifications

Melting Point 108°C
Quantity 25 g
Formula Weight 301.97
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Dibromo-2-naphthol
RTECSNumber QL3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.