Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Dibromohexane 97.0+%, TCI America™
SDP

Catalog No. D0185100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0185100G 100 g
D018525G 25 g
D0185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0185100G Supplier TCI America Supplier No. D0185100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 246°C
Quantity 100 g
Formula Weight 243.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dibromohexane
RTECSNumber MO1515000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.