Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diiodohexane 99+%, Thermo Scientific™

Catalog No. AC243210500
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC243210500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243210500 Supplier Thermo Scientific Chemicals Supplier No. 243210500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 629-09-4
CAS Min % 99.0
CAS Max % 100.0
Density 2.052
Infrared Spectrum Authentic
Molecular Formula C6H12I2
Refractive Index 1.5852 to 1.5872 (20°C, 589nm)
Linear Formula I(CH2)6I
Quantity 50g
MDL Number MFCD00001103
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
SMILES ICCCCCCI
IUPAC Name 1,6-diiodohexane
Molecular Weight (g/mol) 337.97
PubChem CID 12373
Formula Weight 337.96
Percent Purity ≥99.0%
Chemical Name or Material 1, 6-Diiodohexane, 99+%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 211-073-5
RTECSNumber MO1700000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.