Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,6-Dimethylnaphthalene

Catalog No. 250937250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-093-7250MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-093-7250MG Supplier Sigma Aldrich Supplier No. 250937250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,6-Dimethylnaphthalene

Specifications

Melting Point -17°C to +16°C (lit.)
CAS 575-43-9
Boiling Point 265°C to 266°C (lit.)
Molecular Formula C12H12
Refractive Index n20/D 1.606 (literature)
Linear Formula C10H6(CH3)2
MDL Number MFCD00004039
Density 1.002 g/mL (at 25°C (literature))
Molecular Weight (g/mol) 156.22
Quantity 100 mg
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.