Learn More
Medchemexpress LLC 1,6-heptadiene-3,5-dione, 1,7-bis(4-hydroxy-3-methoxyphenyl)-, (1E,6E)- | 458-37-7 | MFCD00008365 | 97.3% | 368.38 g·mol^-1 | C21H20O6 | 10 MG
SDP
Catalog No. 5000298985
Encompass_Preferred
Curcumin (diferuloylmethane) is a natural phenolic research reagent that functions as a p300/CREB-binding protein-specific inhibitor of histone acetyltransferase. It modulates NF-κB and MAPK pathways, stabilizes Nrf2 through Keap1 cysteine modification, and exhibits anti-inflammatory, antioxidant, antiproliferative, and antiangiogenic activities. Supplied as a characterized powder with recommended storage conditions and a COA.
Catalog No. 50-002-98985 Supplier Medchemexpress LLC Supplier No. HYN0005R10MG
Curcumin (diferuloylmethane) is a natural phenolic research reagent that functions as a p300/CREB-binding protein-specific inhibitor of histone acetyltransferase. It modulates NF-κB and MAPK pathways, stabilizes Nrf2 through Keap1 cysteine modification, and exhibits anti-inflammatory, antioxidant, antiproliferative, and antiangiogenic activities. Supplied as a characterized powder with recommended storage conditions and a COA.