Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Heptadiene 99%, Thermo Scientific™

Catalog No. AC338350010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338350010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338350010 Supplier Thermo Scientific Chemicals Supplier No. 338350010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3070-53-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00008667
InChI Key GEAWFZNTIFJMHR-UHFFFAOYSA-N
Synonym 1,6-heptadiene, ch2=ch ch2 3ch=ch2, acmc-1cs8a
PubChem CID 16968
IUPAC Name hepta-1,6-diene
SMILES C=CCCCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.71
Infrared Spectrum Authentic
Refractive Index 1.4135 to 1.4155 (20°C, 589nm)
Linear Formula CH2CH(CH2)3CHCH2
Quantity 1 g
Formula Weight 96.17
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1, 6-Heptadiene, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.