Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ 1,6-Hexamethylenediamine, 0.5M Solution
SCI_ED

Catalog No. S25349 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
0.5M
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25349 250 mL 0.5M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25349
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 102601-60-5 , 124-9-4 , 7732-18-5
Molecular Formula C34H32ClCoN4O4
Molecular Weight (g/mol) 655.037
InChI Key WWNXKQFLDNSNDN-UHFFFAOYSA-K
Synonym cobaltic protoporphyrin ix chloride, 3-18-2-carboxyethyl-7,12-bis ethenyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid;chlorocobalt 2+ \n3-18-2-carboxyethyl-7,12-bis ethenyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid;chlorocobalt 2+, 3-18-2-carboxyethyl-8,13-bis ethenyl-3,7,12,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; chlorocobalt 2+
PubChem CID 50910263
IUPAC Name 3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;chlorocobalt(2+)
SMILES CC1=C(C2=CC3=C(C(=C([N-]3)C=C4C(=C(C(=N4)C=C5C(=C(C(=N5)C=C1[N-]2)C=C)C)C=C)C)C)CCC(=O)O)CCC(=O)O.Cl[Co+2]

Specifications

Density 1g/mL
Assay Percent Range <2.0% v/v
Packaging 0.5M
Linear Formula H2N(CH2)6NH2
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material 1, 6-Hexamethyldiamine, 0.5M Solution
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.