Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanediamine, 99.5+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
124-09-4
C6H16N2
116.21
NAQMVNRVTILPCV-UHFFFAOYSA-N
1,6-hexanediamine,1,6-diaminohexane,hexamethylenediamine,hmda,1,6-hexylenediamine,1,6-hexamethylenediamine,1,6-diamino-n-hexane,hexamethylene diamine,hexylenediamine,1,6-hexanediamine solution
16402
CHEBI:39618
hexane-1,6-diamine
C(CCCN)CCN
124-09-4
C6H16N2
116.21
NAQMVNRVTILPCV-UHFFFAOYSA-N
1,6-hexanediamine,1,6-diaminohexane,hexamethylenediamine,hmda,1,6-hexylenediamine,1,6-hexamethylenediamine,1,6-diamino-n-hexane,hexamethylene diamine,hexylenediamine,1,6-hexanediamine solution
16402
CHEBI:39618
hexane-1,6-diamine
C(CCCN)CCN
Melting Point 38.0°C to 41.0°C
Boiling Point 204.0°C to 205.0°C
Packaging Plastic bottle
Color White
Infrared Spectrum Authentic
Quantity 1kg
Flash Point 81°C
Water 0.05% Max. (K.F.)
Assay Percent Range 99.5+%
Linear Formula H2N(CH2)6NH2
Beilstein 04,269
Merck Index 15,4727
Solubility Information Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether
Formula Weight 116.21
Physical Form Low Melting Crystalline Mass
Percent Purity ≥99.5%
Chemical Name or Material 1, 6-Hexanediamine
Show More Show Less