Learn More
Medchemexpress LLC 1,6-Hexanediaminium, N1,N1,N1,N6,N6,N6-hexamethyl-, bromide (1:2) | 55-97-0 | 99.9% | 25 G
SDP
Catalog No. 5000406579
Encompass_Preferred
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) with mixed competitive and noncompetitive activity. It possesses anti-hypertensive properties and has been shown to attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Non-selective ganglionic nicotinic-receptor antagonist (nAChR)
  • Exhibits mixed competitive and noncompetitive activity
  • Attenuates sympathetic activity and blood pressure
  • Targets apoptosis, cardiovascular disease, and neurological disease
  • Appears as a white to off-white solid
Catalog No. 50-004-06579 Supplier Medchemexpress LLC Supplier No. 25GHYB0569
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) with mixed competitive and noncompetitive activity. It possesses anti-hypertensive properties and has been shown to attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Non-selective ganglionic nicotinic-receptor antagonist (nAChR)
  • Exhibits mixed competitive and noncompetitive activity
  • Attenuates sympathetic activity and blood pressure
  • Targets apoptosis, cardiovascular disease, and neurological disease
  • Appears as a white to off-white solid