Learn More
Medchemexpress LLC 1,6-Hexanediol | 629-11-8 | 99.9% | 118.17 | 500 G
SDP
Catalog No. 5000453556
Encompass_Preferred
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be utilized as an additive for drilling fluids and as a substrate in life science-related research.
  • 6-carbon straight-chain diol with hydroxyl groups at both ends.
  • Can be used as an additive for drilling fluids.
  • Can be used as a substrate in life science-related research.
  • Its monoesters and diesters act as substrates for in vitro digestion studies by pancreatic lipase.
  • Can be catalytically lactonized by alcohol dehydrogenases to produce ε-caprolactone.
Catalog No. 50-004-53556 Supplier Medchemexpress LLC Supplier No. HYW010505500G
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be utilized as an additive for drilling fluids and as a substrate in life science-related research.
  • 6-carbon straight-chain diol with hydroxyl groups at both ends.
  • Can be used as an additive for drilling fluids.
  • Can be used as a substrate in life science-related research.
  • Its monoesters and diesters act as substrates for in vitro digestion studies by pancreatic lipase.
  • Can be catalytically lactonized by alcohol dehydrogenases to produce ε-caprolactone.