Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanediol Dimethacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. H1369500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1369500G 500 g
H136925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1369500G Supplier TCI America Supplier No. H1369500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6606-59-3
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.326
MDL Number MFCD00015425
InChI Key SAPGBCWOQLHKKZ-UHFFFAOYSA-N
Synonym 1,6-hexanediol dimethacrylate, unii-rao49190jv, 1,6-hexanediyl bismethacrylate, ccris 7049, 1,6-hexamethylene dimethacrylate, 2-propenoic acid, 2-methyl-, 1,6-hexanediyl ester, hexane-1,6-diyl bis 2-methylacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,6-hexanediyl ester, 1,6-hexanediyl bis 2-methylacrylate, 6-2-methylprop-2-enoyl oxy hexyl 2-methylprop-2-enoate
PubChem CID 81080
IUPAC Name 6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C

Specifications

Color Yellow
Quantity 500 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Hexanediol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1059
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.