Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanedithiol, 97%

Catalog No. AC120670250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120670250 25 g Glass bottle
AC120670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120670250 Supplier Thermo Scientific Chemicals Supplier No. 120670250
  • $172.30
    $146.65 / Each of 1

    Save $25.65

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-43-1
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00004910
InChI Key SRZXCOWFGPICGA-UHFFFAOYSA-N
Synonym 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
PubChem CID 14491
IUPAC Name hexane-1,6-dithiol
SMILES SCCCCCCS

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -21.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 237.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5110 to 1.5150
Linear Formula HS(CH2)6SH
Quantity 25 g
Beilstein 01, 484
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in fat
Formula Weight 150.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 6-Hexanedithiol, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-735-1
RTECSNumber MO3500000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.