Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Naphthyridine 97.0+%, TCI America™
SDP

Catalog No. N04621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04621G 1 g
N0462100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04621G Supplier TCI America Supplier No. N04621G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 253-72-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00059750
InChI Key VSOSXKMEQPYESP-UHFFFAOYSA-N
Synonym 1,6-diazanaphthalene, 1,6 naphthyridine, 1,6-pyridopyridine, unii-s1t14p7ow7, pyridino 4,3-b pyridine, 1,6naphthyridine, 1,6-naphthyridine, pubchem18071, acmc-1chvm, pyrido 4,3-b pyridine
PubChem CID 67488
ChEBI CHEBI:36627
IUPAC Name 1,6-naphthyridine
SMILES C1=CN=C2C=CN=CC2=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 130.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Naphthyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.