Learn More
Cayman Chemical 1 6-nhydro-D-galactose 100mg
SDP
Catalog No. 502983246
Encompass_Preferred
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter
Catalog No. 50-298-3246 Supplier Cayman Chemical Supplier No. 18895100 MG
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter