Learn More
Cayman Chemical 1,6-Anhydro-D-galactose, Purity: ≥95%, CAS: 644-76-8, 1 g
SDP
Catalog No. 502982794
Encompass_Preferred
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter
Catalog No. 50-298-2794 Supplier Cayman Chemical Supplier No. 188951 G
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter