Learn More
Cayman Chemical 1 6-nhydro-D-galactose 250mg
SDP
Catalog No. 502981899
Encompass_Preferred
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter
Catalog No. 50-298-1899 Supplier Cayman Chemical Supplier No. 18895250 MG
Add to Cart
Add to Cart
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter