Learn More
Cayman Chemical 1,6-Anhydro-D-galactose, Purity: ≥95%, CAS: 644-76-8, 500 mg
SDP
Catalog No. 502984182
Encompass_Preferred
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter
Catalog No. 50-298-4182 Supplier Cayman Chemical Supplier No. 18895500 MG
A carbohydrate found in liquid smoke flavorings that is used as a tracer of the contribution of biomass burning to total atmospheric particulate matter