Learn More
Medchemexpress LLC 1,6-Octadien-3-ol, 3,7-dimethyl- | 78-70-6 | 154.25 | 10 G
SDP
Catalog No. 5000389843
Encompass_Preferred
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier. It possesses various biological activities.
  • Acts as a competitive NMDA receptor antagonist
  • Orally active and can cross the blood-brain barrier
  • Exhibits anticancer activity
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotective activity
  • Exhibits anxiolytic activity
  • Exhibits antidepressant activity
  • Exhibits anti-stress activity
  • Exhibits cardioprotective activity
  • Exhibits hepatoprotective activity
  • Exhibits nephroprotective activity
  • Exhibits pulmonary protective activity
Catalog No. 50-003-89843 Supplier Medchemexpress LLC Supplier No. HYN036810G
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier. It possesses various biological activities.
  • Acts as a competitive NMDA receptor antagonist
  • Orally active and can cross the blood-brain barrier
  • Exhibits anticancer activity
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotective activity
  • Exhibits anxiolytic activity
  • Exhibits antidepressant activity
  • Exhibits anti-stress activity
  • Exhibits cardioprotective activity
  • Exhibits hepatoprotective activity
  • Exhibits nephroprotective activity
  • Exhibits pulmonary protective activity