Learn More
Medchemexpress LLC 1,6-Octadien-3-ol, 3,7-dimethyl- | 78-70-6 | 154.25 | 25 G
SDP
Catalog No. 5000389331
Encompass_Preferred
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier. This compound possesses various activities including anticancer, antibacterial, anti-inflammatory, neuroprotective, anxiolytic, antidepressant, anti-stress, cardioprotective, hepatoprotective, nephroprotective, and pulmonary protective properties.
  • Acts as a competitive NMDA receptor antagonist.
  • Orally active and can cross the blood-brain barrier.
  • Exhibits anticancer activity.
  • Exhibits antibacterial activity.
  • Possesses anti-inflammatory activity.
  • Offers neuroprotective benefits.
  • Has anxiolytic and antidepressant effects.
  • Provides anti-stress, cardioprotective, hepatoprotective, nephroprotective, and pulmonary protective activities.
Catalog No. 50-003-89331 Supplier Medchemexpress LLC Supplier No. 25GHYN0368
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier. This compound possesses various activities including anticancer, antibacterial, anti-inflammatory, neuroprotective, anxiolytic, antidepressant, anti-stress, cardioprotective, hepatoprotective, nephroprotective, and pulmonary protective properties.
  • Acts as a competitive NMDA receptor antagonist.
  • Orally active and can cross the blood-brain barrier.
  • Exhibits anticancer activity.
  • Exhibits antibacterial activity.
  • Possesses anti-inflammatory activity.
  • Offers neuroprotective benefits.
  • Has anxiolytic and antidepressant effects.
  • Provides anti-stress, cardioprotective, hepatoprotective, nephroprotective, and pulmonary protective activities.