Learn More
Medchemexpress LLC 1,6-Octadien-3-ol, 3,7-dimethyl- | 78-70-6 | 154.25 | 50 G
SDP
Catalog No. 5000390018
Encompass_Preferred
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier.
  • Anticancer activity
  • Antibacterial activity
  • Anti-inflammatory activity
  • Neuroprotective activity
  • Anxiolytic activity
  • Antidepressant activity
  • Anti-stress activity
  • Cardioprotective activity
  • Hepatoprotective activity
  • Nephroprotective activity
  • Pulmonary protective activity
Catalog No. 50-003-90018 Supplier Medchemexpress LLC Supplier No. HYN036850G
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier.
  • Anticancer activity
  • Antibacterial activity
  • Anti-inflammatory activity
  • Neuroprotective activity
  • Anxiolytic activity
  • Antidepressant activity
  • Anti-stress activity
  • Cardioprotective activity
  • Hepatoprotective activity
  • Nephroprotective activity
  • Pulmonary protective activity