Learn More
Thermco Products Inc Non-Mercury Fractional Degree Thermometer - Red Spirit | -1 to 61 °C
Catalog No. NC1810152
Encompass_Preferred
Non-Mercury Fractional Degree Thermometer has a range of -1 to 61 °C with 0.1°C divisions and a 420mm length.
Catalog No. NC1810152 Supplier Thermco Products Inc Supplier No. ACC10613S
Non-Mercury Fractional Degree Thermometer has a range of -1 to 61 °C with 0.1°C divisions and a 420mm length.