Learn More
Andwin Scientific Laser Cryo-Tags, 1.69 x 0.75", Rainbow Pack, 1,040/pk
SDP
Catalog No. NC1662627
Encompass_Preferred
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.
Catalog No. NC1662627 Supplier Andwin Scientific Supplier No. RNBW2000
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.