Learn More
Next Advance Inc 1.6MM SS BALLS 1LB
SDP
Catalog No. NC9957299
Encompass
1.6 mm Stainless steel beads, density is 7.9 g/cc, 1 lb (.45 kg). For the homogenization of tough tissue and cells. Approximately 33,000 beads in a package.
Catalog No. NC9957299 Supplier Next Advance Inc Supplier No. SSB16
1.6 mm Stainless steel beads, density is 7.9 g/cc, 1 lb (.45 kg). For the homogenization of tough tissue and cells. Approximately 33,000 beads in a package.