Learn More
Medchemexpress LLC 1-(7-Bromobenzo[d]dioxol-4-yl)ethan-1-one | 1892297-27-6 | 243.06 | 250 MG
SDP
Catalog No. 5000402160
Encompass_Preferred
1-(7-Bromobenzo[d]dioxol-4-yl)ethan-1-one is a drug intermediate used for the synthesis of various active compounds.
  • Drug intermediate
  • Used for synthesis of various active compounds
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-02160 Supplier Medchemexpress LLC Supplier No. 250MGHY42238
1-(7-Bromobenzo[d]dioxol-4-yl)ethan-1-one is a drug intermediate used for the synthesis of various active compounds.
  • Drug intermediate
  • Used for synthesis of various active compounds
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month