Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Dichloroheptane 90.0+%, TCI America™
SDP

Catalog No. D10305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10305G Supplier TCI America Supplier No. D10305G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 821-76-1
Molecular Formula C7H14Cl2
Molecular Weight (g/mol) 169.09
MDL Number MFCD00049144
InChI Key PSEMXLIZFGUOGC-UHFFFAOYSA-N
Synonym Heptamethylene Chloride, Heptamethylene Dichloride
PubChem CID 69965
IUPAC Name 1,7-dichloroheptane
SMILES ClCCCCCCCCl

Specifications

Color Colorless
Quantity 5 g
UN Number 3082
Formula Weight 169.09
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Dichloroheptane
EINECSNumber (2)-0066
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.