Learn More
AdipoGen 1 7-Dimethyluric acid
SDP
Catalog No. 502561143
Encompass_Preferred
Chemical. CAS 33868-03-0. Formula C7H8N4O3. MW 196.2. Synthetic. A metabolite of caffeine with antioxidant and neuroprotective activities. Does not inhibit the differentiation of 3T3-L1 pre-adipocytes to mature adipocytes, but suppress the intracellular lipid accumulation after complete differentiation in a dose-dependent manner.
Catalog No. 50-256-1143 Supplier AdipoGen Supplier No. CDXD0295M050
Chemical. CAS 33868-03-0. Formula C7H8N4O3. MW 196.2. Synthetic. A metabolite of caffeine with antioxidant and neuroprotective activities. Does not inhibit the differentiation of 3T3-L1 pre-adipocytes to mature adipocytes, but suppress the intracellular lipid accumulation after complete differentiation in a dose-dependent manner.