Learn More
Cayman Chemical 1,7-Dimethyluric Acid, Purity: ≥98%, CAS: 33868-03-0, 50 mg
SDP
Catalog No. 502981722
Encompass_Preferred
A metabolite of caffeine that can be detected in urine as a biomarker of caffeine consumption
Catalog No. 50-298-1722 Supplier Cayman Chemical Supplier No. 1958450 MG
A metabolite of caffeine that can be detected in urine as a biomarker of caffeine consumption