Learn More
Medchemexpress LLC 1,7-Heptanediamine | 646-19-5 | 98.54% | 130.23 | 10 G
SDP
Catalog No. 5000452133
Encompass_Preferred
1,7-Heptanediamine (C7) is a deoxyhypusine synthase inhibitor with an IC50 value of 4.5 μM. It appears as a solid below 26°C and a liquid above 29°C, with a colorless to off-white appearance. It is soluble in water at ≥ 100 mg/mL (767.87 mM).
  • Deoxyhypusine synthase inhibitor with an IC50 value of 4.5 μM
  • Appears as a solid below 26°C and a liquid above 29°C
  • Colorless to off-white appearance
  • Soluble in water at ≥ 100 mg/mL
Catalog No. 50-004-52133 Supplier Medchemexpress LLC Supplier No. HYW00882110G
1,7-Heptanediamine (C7) is a deoxyhypusine synthase inhibitor with an IC50 value of 4.5 μM. It appears as a solid below 26°C and a liquid above 29°C, with a colorless to off-white appearance. It is soluble in water at ≥ 100 mg/mL (767.87 mM).
  • Deoxyhypusine synthase inhibitor with an IC50 value of 4.5 μM
  • Appears as a solid below 26°C and a liquid above 29°C
  • Colorless to off-white appearance
  • Soluble in water at ≥ 100 mg/mL