Learn More
Laxco™ 1.7 Megapixel High End Digital CMOS Camera
GREENER_CHOICESDP
Catalog No. 12563683 Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Catalog No. Voltage
12-563-683 100/115/220/230 V
1 options
Catalog No. 12-563-683 Supplier Laxco™ Supplier No. SEBACAMCOOL1.7C
Add to Cart
Add to Cart
Request a Quote
Cooled color camera for high resolution imaging and timelapse applications. Best suited for your fluorescence imaging needs.
  • 1.7 megapixels cooled camera
  • Full live color images/videos
  • Best camera for fluorescence imaging
  • Works best with upright and inverted microscope
  • Needs c-mount to attach with microscope
Specifications
Type Digital Camera
Autoclavable Not Autoclavable
For Use With (Equipment) SEBAP4C2 Microscope
Sterility Sterile
Voltage 100/115/220/230 V