Learn More
Andwin Scientific 1.7 ML GRDTD MICROTUBE 250/PK
SDP
Catalog No. NC9411672
Encompass_Preferred
The graduated microtube features low adhesion and clear body that makes this product great for easy removal of DNA pellets. It is ideal for use for DNA sequencing or any application where a lubricated tube is required. The microtube has a volume capacity of 1.7 mL. This is sold in a pack of 250.
Catalog No. NC9411672 Supplier Andwin Scientific Supplier No. C33021
The graduated microtube features low adhesion and clear body that makes this product great for easy removal of DNA pellets. It is ideal for use for DNA sequencing or any application where a lubricated tube is required. The microtube has a volume capacity of 1.7 mL. This is sold in a pack of 250.