Learn More
Andwin Scientific 1.7 SNAP CAP VIALS 500/PK
SDP
Catalog No. NC9819718
Encompass_Preferred
The GeneMate 1.7 mL Graduated Microcentrifuge Tubes are ideal for boiling applications. The tubes work within the temperature range of -90C to 120C. This product features wide cap flange that permits easy opening. The tubes are superior in clarity and are free of detectable DNase, RNase, and human DNA. It is sold in a pack of 500 tubes in clear color.
Catalog No. NC9819718 Supplier Andwin Scientific Supplier No. C32621
The GeneMate 1.7 mL Graduated Microcentrifuge Tubes are ideal for boiling applications. The tubes work within the temperature range of -90C to 120C. This product features wide cap flange that permits easy opening. The tubes are superior in clarity and are free of detectable DNase, RNase, and human DNA. It is sold in a pack of 500 tubes in clear color.