Learn More
Medchemexpress LLC 1-7-Substance P, 2,2,2-trifluoroacetate (1:1) | 2828433-22-1 | 99.1% | 1014.06 | 10 MG
SDP
Catalog No. 5000423287
Encompass_Preferred
Substance P(1-7) TFA is a fragment of the neuropeptide, substance P (SP). It gives depressor and bradycardic effects when applied to the nucleus tractus solitarius. In vivo, it acts as a potent antagonist against SP-induced responses and is proposed as an endogenous modulator of SP actions. Injection of low doses can reduce aversive behaviors induced by SP or SP(5-11).
  • Fragment of the neuropeptide substance P (SP)
  • Gives depressor and bradycardic effects
  • Acts as a potent antagonist against SP-induced responses
  • Proposed as an endogenous modulator of SP actions
  • Reduces aversive behaviors
  • White to off-white solid appearance
  • Sequence: Arg-Pro-Lys-Pro-Gln-Qln-Phe
Catalog No. 50-004-23287 Supplier Medchemexpress LLC Supplier No. HYP1485A10MG
Substance P(1-7) TFA is a fragment of the neuropeptide, substance P (SP). It gives depressor and bradycardic effects when applied to the nucleus tractus solitarius. In vivo, it acts as a potent antagonist against SP-induced responses and is proposed as an endogenous modulator of SP actions. Injection of low doses can reduce aversive behaviors induced by SP or SP(5-11).
  • Fragment of the neuropeptide substance P (SP)
  • Gives depressor and bradycardic effects
  • Acts as a potent antagonist against SP-induced responses
  • Proposed as an endogenous modulator of SP actions
  • Reduces aversive behaviors
  • White to off-white solid appearance
  • Sequence: Arg-Pro-Lys-Pro-Gln-Qln-Phe