Learn More
Bellco Glass, Inc. 1.7in. Probe Holder,45mm
SDP
Catalog No. 501944940
Encompass_Preferred
Probe adapters & compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. 13.5 thread for pH & DO probes. MADE IN THE USA.
Catalog No. 50-194-4940 Supplier Bellco Glass, Inc. Supplier No. 196491015
Probe adapters & compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. 13.5 thread for pH & DO probes. MADE IN THE USA.