Learn More
Alkali Scientific Qwik-Snap™1.7mL Sterile Microtube, Packed in Self-standing Bags of 50, 10 Sterile Inner Bags, Clear, 500/cs
SDP
Catalog No. NC2022341
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 clear, sterile 1.7mL centrifuge tubes.
Catalog No. NC2022341 Supplier Alkali Scientific Supplier No. C3017ST
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 clear, sterile 1.7mL centrifuge tubes.