Learn More
Alkali Scientific Qwik-Snap™ 1.7mL Clear Microcentrifuge Tubes, Sterile, 500/pk
SDP
Catalog No. NC1377901
Encompass_Preferred
Alkali Scientific Qwik-Snap™ 1.7mL MicroTubes are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Certified RNase, DNase and pyrogen free. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.
Catalog No. NC1377901 Supplier Alkali Scientific Supplier No. C3016S
Alkali Scientific Qwik-Snap™ 1.7mL MicroTubes are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Certified RNase, DNase and pyrogen free. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.