Learn More
Biotang Inc 1.7 ml Rainbow Pack Microcentrifuge Tubes. 500/bag, 5000/ 10bag/case. Superior clarity. RCF rating of 25,000 x g. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.
Catalog No. 507515025 Shop All BIOTANG Inc Products
Encompass_Preferred
1.7 ml Rainbow Pack Microcentrifuge Tubes. 500/bag, 5000/ 10bag/case. Superior clarity. RCF rating of 25,000 x g. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.
Catalog No. 50-751-5025 Supplier Biotang Inc Supplier No. BT11590
1.7 ml Rainbow Pack Microcentrifuge Tubes. 500/bag, 5000/ 10bag/case. Superior clarity. RCF rating of 25,000 x g. Fit all standard rotors.Autoclavable. Dnase, Rnase, Human DNA, Pyrogen, PCR hibitors free.