Learn More
Alkali Scientific Qwik-Snap™ 1.7mL Microcentrifuge Tubes, Purple, 500/pk
SDP
Catalog No. 502388747
Encompass_Preferred
Alkali Scientific Qwik-Snap™ 1.7mL MicroTubes are made of premium purple polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.
Catalog No. 50-238-8747 Supplier Alkali Scientific Supplier No. C3017V
Alkali Scientific Qwik-Snap™ 1.7mL MicroTubes are made of premium purple polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.