Learn More
Genesee Scientific Corporation Genesee Scientific 1.7 mL Clear Microtubes, Certified Free of RNase/DNase, Autoclavable, 500 Tubes
SDP
Catalog No. NC1940918
Encompass
The Genesee Scientific 1.7 mL Clear Microtubes are high-quality polypropylene tubes designed for diverse scientific applications. Featuring a positive snap seal cap for secure closure, these tubes are certified free from RNase, DNase, and endotoxins, ensuring sample integrity. They are autoclavable, boilproof, and can withstand extreme temperatures, making them suitable for demanding research environments. With a glass-like clarity and frosted writing surface, they provide excellent sample identification and handling.
Catalog No. NC1940918 Supplier Genesee Scientific Corporation Supplier No. 24282
Add to Cart
Add to Cart
The Genesee Scientific 1.7 mL Clear Microtubes are high-quality polypropylene tubes designed for diverse scientific applications. Featuring a positive snap seal cap for secure closure, these tubes are certified free from RNase, DNase, and endotoxins, ensuring sample integrity. They are autoclavable, boilproof, and can withstand extreme temperatures, making them suitable for demanding research environments. With a glass-like clarity and frosted writing surface, they provide excellent sample identification and handling.