Learn More
Genesee Scientific Corporation 1.7ML MICROTUBES,YELLOW,500/CS
SDP
Catalog No. NC2045332
Encompass
Olympus 1.7ml Microtubes, YellowPolypropylene, BoilproofBox of 500 Tubes/UnitBrand: Olympus PlasticsCertified free from RNase, DNase, DNA & PCR inhibitors. Also Endotoxin-FreeAutoclavable, Temp Range: -90ºC to 121ºCRated to >20,000 x gGraduations every 100µl - easier estimation of contents!Frosted side writing surfaceExceptionally clear - glass-like clarityBoilproof protection w/ Positive SealMade in the USA
Catalog No. NC2045332 Supplier Genesee Scientific Corporation Supplier No. 24282Y
Olympus 1.7ml Microtubes, YellowPolypropylene, BoilproofBox of 500 Tubes/UnitBrand: Olympus PlasticsCertified free from RNase, DNase, DNA & PCR inhibitors. Also Endotoxin-FreeAutoclavable, Temp Range: -90ºC to 121ºCRated to >20,000 x gGraduations every 100µl - easier estimation of contents!Frosted side writing surfaceExceptionally clear - glass-like clarityBoilproof protection w/ Positive SealMade in the USA