Learn More
Andwin Scientific 1.7ML SAFE SEAL CLR MCT 500/PK
SDP
Catalog No. NC9730030
Encompass_Preferred
The MlTI SafeSeal Microcentrifuge Tubes are disposable tubes made for biotechnology applications. This product is made from precision molded polypropylene. Caps are attached onto the tubes and are specially designed to allow users to easily open the tubes, but at the same time ensure secure sample storage during boiling procedures. This product has a volume capacity of 1.7 mL and is sold in a pack of 500 natural-colored tubes.
Catalog No. NC9730030 Supplier Andwin Scientific Supplier No. 53550698
The MlTI SafeSeal Microcentrifuge Tubes are disposable tubes made for biotechnology applications. This product is made from precision molded polypropylene. Caps are attached onto the tubes and are specially designed to allow users to easily open the tubes, but at the same time ensure secure sample storage during boiling procedures. This product has a volume capacity of 1.7 mL and is sold in a pack of 500 natural-colored tubes.