Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8,9-Trihydroxyanthracene 95.0+%, TCI America™
SDP

Catalog No. T116610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T116610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T116610G Supplier TCI America Supplier No. T116610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 480-22-8
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00001250
InChI Key YUTJCNNFTOIOGT-UHFFFAOYSA-N
Synonym Anthralin, Dithranol, 1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, DIT
PubChem CID 10187
ChEBI CHEBI:2756
IUPAC Name anthracene-1,8,9-triol
SMILES C1=CC2=CC3=C(C(=CC=C3)O)C(=C2C(=C1)O)O

Specifications

Melting Point 181°C
Quantity 10 g
Formula Weight 226.23
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8,9-Trihydroxyanthracene
RTECSNumber CB1225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.