Learn More
AVANTOR MASTERFLEX (VWR LLC) BarbLock Assembly Hand Tool for 1/8"" tubing
Catalog No. NC0924813
Encompass
A PVDF and stainless steel assembly hand tool for 1/8" tubing. Select a hand tool to assemble your BarbLock Retainers based on the tubing size. A universal removal tool also is available.
Catalog No. NC0924813 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3060418
A PVDF and stainless steel assembly hand tool for 1/8" tubing. Select a hand tool to assemble your BarbLock Retainers based on the tubing size. A universal removal tool also is available.