Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(2-pyridyl)-3,6-dithiaoctane 98.0+%, TCI America™
SDP

Catalog No. B12295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12295G 5 g
B12291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12295G Supplier TCI America Supplier No. B12295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64691-70-9
Molecular Formula C16H20N2S2
Molecular Weight (g/mol) 304.47
MDL Number MFCD00006366
InChI Key YDPDVMHQFYTZDI-UHFFFAOYSA-N
PubChem CID 300754
IUPAC Name 2-[2-[2-(2-pyridin-2-ylethylsulfanyl)ethylsulfanyl]ethyl]pyridine
SMILES C1=CC=NC(=C1)CCSCCSCCC2=CC=CC=N2

Specifications

Melting Point 51°C
Quantity 5 g
Synonym S,S′-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2′-(3,6-Dithiaoctamethylene)pyridine
Formula Weight 304.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(2-pyridyl)-3,6-dithiaoctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.