Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(diphenylphosphino)naphthalene 97.0+%, TCI America™
SDP

Catalog No. B45301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45301G 1 g
B45305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45301G Supplier TCI America Supplier No. B45301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153725-04-3
Molecular Formula C34H26P2
Molecular Weight (g/mol) 496.53
MDL Number MFCD04117326
InChI Key QOXZKPURCFVBRR-UHFFFAOYSA-N
PubChem CID 2756729
IUPAC Name [8-(diphenylphosphanyl)naphthalen-1-yl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC2=CC=CC(P(C3=CC=CC=C3)C3=CC=CC=C3)=C12

Specifications

Melting Point 210°C
Color Yellow
Quantity 1 g
Formula Weight 496.53
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(diphenylphosphino)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.