1,8-Cineole, 99%

Manufacturer: Alfa Aesar A12269AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA12269AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 470-82-6
    Molecular Formula C10H18O
    Formula Weight 154.24
    MDL Number MFCD00167977
    UN Number UN3271
    Beilstein 105109
    Merck Index 14,3895
    Chemical Name or Material Cineole

    Specifications

    Formula Weight 154.24
    Quantity 100mL
    TSCA TSCA
    MDL Number MFCD00167977
    UN Number UN3271
    Beilstein 105109
    Merck Index 14,3895
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    RTECS Number OS9275000
    Physical Form Liquid
    Density 0.92
    Assay Percent Range 99%
    CAS 470-82-6
    Molecular Formula C10H18O
    Chemical Name or Material Cineole
    Assay 99%

    Description

    Applications
    Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

    Solubility
    Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.

    Notes
    Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit