Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Cineole 99.0+%, TCI America™
SDP

Catalog No. C054225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C054225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C054225ML Supplier TCI America Supplier No. C054225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 2°C
Color Colorless
Boiling Point 176°C
Quantity 25 mL
UN Number 1993
Formula Weight 154.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
EINECSNumber (5)-0684
RTECSNumber OS9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.